Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate, Monohydrate, Granular, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605530 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
50 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-530 2.5 kg Poly Bottle
18-605-532 50 g Poly Bottle
18-605-529 500 g Poly Bottle
18-605-531 12 kg Poly Pail
18-605-533 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-530 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO1752.5KGPL
Only null left
Add to Cart
Add to Cart

PO175, 6100-05-6, C6H5K3O7·H2O

Spectrum™ Chemical Potassium Citrate, Monohydrate, Granular, USP belongs to a class of drugs known as urinary alkalinizers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 3-6%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.