Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagents

Catalog No. 01337856
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-856 2.5 kg
01-337-857 125 g
01-337-855 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-856 Supplier Reagents Holdings Llc Supplier No. C12815002.5A5
Add to Cart
Add to Cart

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym Tripotassium Citrate Monohydrate
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Boiling Point 230°C
Melting Point 180°C (anh)
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.98
Chemical Name or Material Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.