Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.200 N (N/5), Ricca Chemical
SDP

Catalog No. 60651 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6065-1 4 L
606516 500 mL
6065-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6065-1 Supplier Ricca Chemical Company Supplier No. 60651
Add to Cart
Add to Cart

Specifications

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
Molecular Weight (g/mol) 294.18 g/mol
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.200N (N/5)
Packaging Natural Poly Bottle
pH ∼4
Quantity 4 L
Chemical Name or Material Potassium Dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.