Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), Reagents

Catalog No. 01337846
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-846 500 g
01-337-849 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-846 Supplier Reagents Holdings Llc Supplier No. C1283000500C2
Add to Cart
Add to Cart

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym Potassium Hexacyanoferrate III, Red Prussiate, Potassium Ferricyanate
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 300°C
Color Orange/Red
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.89
Chemical Name or Material Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III))
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.