Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferricyanide, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605542 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-542 500 g Amber Glass Bottle
18-605-539 25 g Amber Glass Bottle
18-605-540 125 g Amber Glass Bottle
18-605-541 6 x 125 g Amber Glass Bottle
18-605-544 2.5 kg Amber Glass Bottle
18-605-543 6 x 500 g Amber Glass Bottle
18-605-545 4 x 2.5 kg Amber Glass Bottle
18-605-546 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-605-542 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1285500GM
Add to Cart
Add to Cart

P1285, 13746-66-2, K3Fe(CN)6

Spectrum™ Chemical Potassium Ferricyanide, Crystal, Reagent, ACS is a chemical compound and a bright red salt. It is soluble in water and appears as a yellow-green fluorescence. Potassium ferricyanide is created by passing chlorine through a solution of potassium ferrocyanide. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.