Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 30% (w/w), Ricca Chemical
SDP

Catalog No. 61035 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61035 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61035 Supplier Ricca Chemical Company Supplier No. 61035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color Colorless to Pink
CAS Min % 69.0, 29.41
CAS Max % 71.0, 30.61
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH ∼7
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 30% (w/w)
Percent Purity 30%
Grade Laboratory
Chemical Name or Material Potassium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.