Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride, 40 wt.% on alumina, Thermo Scientific Chemicals

Catalog No. AC390811000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC390811000 100 g
AC390810250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC390811000 Supplier Thermo Scientific Chemicals Supplier No. 390811000
  • $285.00
    $242.65 / Each of 1

    Save $42.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium;fluoride
SMILES [F-].[K+]

Specifications

Quantity 100 g
Linear Formula KF
Merck Index 15,7753
Formula Weight 58.09
Packaging Glass bottle
Chemical Name or Material Potassium fluoride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 232-151-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.