Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 50% (w/v) Solution, Spectrum™ Chemical
SDP

Catalog No. 18612649 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-649 4 L Cubitainer
18-612-637 500 mL Cubitainer
18-612-648 1 L Cubitainer
18-612-636 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-649 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P4594LTPL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7789-23-3

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5 , 13455-21-5

Specifications

Physical Form Solution
Quantity 4 L
Packaging Cubitainer
Grade Ungraded
Chemical Name or Material Potassium Fluoride, 50 Percent (w/v) Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.