Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 99.0% min., ACS, MilliporeSigma™

Catalog No. MPX14674 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX14674 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX14674 Supplier MilliporeSigma Supplier No. PX14674

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99.9% min.
Linear Formula KF
Formula Weight 58.1
Grade ACS
Vapor Pressure 1.3 hPa (885°C)
Density 2.48g/cm3 (20°C); Bulk Density: 400 kg/m3
Chemical Name or Material Potassium Fluoride, For Analysis
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.