Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Anhydrous, Purified, Reagents

Catalog No. 01337104
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-104 500 g
01-337-106 100 g
01-337-103 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-104 Supplier Reagents Holdings Llc Supplier No. C2284000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Fluoride, Anhydrous, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.481
Chemical Name or Material Potassium Fluoride, Anhydrous, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.