Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Dihydrate, Crystal, Reagent, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18605569 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-569 2.5 kg Poly Bottle
18-605-565 25 g Poly Bottle
18-605-566 125 g Poly Bottle
18-605-567 500 g Poly Bottle
18-605-568 6 x 500 g Poly Bottle
18-605-570 12 kg Poly Pail
18-605-571 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-569 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P13102.5KG
Add to Cart
Add to Cart

P1310, 13455-21-5, KF·2H2O

Spectrum™ Chemical Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 102%
Grade Reagent
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.