Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride Dihydrate, Ricca Chemical
SDP

Catalog No. RDCP046512F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP046512F 12 kg
RDCP0465500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCP046512F Supplier Ricca Chemical Company Supplier No. RDCP046512F4
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 12 kg
Percent Purity min 97%
Grade Reagent
Packaging Fiber Drum
Chemical Name or Material Potassium Fluoride Dihydrate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.