Learn More
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | KF |
| UN Number | UN1812 |
| Beilstein | 3902818 |
| Formula Weight | 58.1g/mol |
| Percent Purity | ≥99% (F) |
| Grade | Not Available |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Potassium fluoride |
Safety and Handling
| Hazard Statement | P261-P280-P301 + P310-P311 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.