Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachlororhenate(IV), 99.9% (metals basis)

Catalog No. p-7080299
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA6211303 1 g
AA6211306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA6211303 Supplier Thermo Scientific Chemicals Supplier No. 06211303
Only null left
Add to Cart

CAS: 16940-97-9 | Cl6K2Re | 477.10 g/mol

Potassium hexachlororhenate is used in preparation of supported rhenium tricarbonyls which catalyze propylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-97-9
Molecular Formula Cl6K2Re
Molecular Weight (g/mol) 477.10
MDL Number MFCD00058746
InChI Key HGCRXIXEXSEJGV-UHFFFAOYSA-H
Synonym potassium hexachlororhenate iv, dipotassium;hexachlororhenium 2-, dipotassium hexachlororheniumdiuide
PubChem CID 10939720
IUPAC Name dipotassium;hexachlororhenium(2-)
SMILES [K+].[K+].Cl[Re--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Yellow to Green
Physical Form Crystalline
Quantity 1 g
Linear Formula K2ReCl6
Solubility Information Soluble in water.
Formula Weight 477.12
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.34 g/mL
Assay (metals basis)
Chemical Name or Material Potassium hexachlororhenate (IV)
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-008-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.