Learn More
Potassium hexacyanoferrate(III), 98+%
Description
Potassium hexacyanoferrate(III) is used in reading palimpsests and old manuscripts, in blueprint drawing and in photography (Cyanotype process). It finds application to harden iron and steel, in electroplating, dyeing wool and as a laboratory reagent. It is a mild oxidizing agent used in organic synthesis. It is associated with potassium hydroxide solution used to formulate Murakami’s etchant, which is useful for metallographers to provide contrast between binder and carbide phases in cemented carbides. Further, it is used in many amperometric biosensors as an electron transfer agent replacing an enzyme's natural electron transfer agent such as oxygen with the enzyme glucose oxidase. It is an active ingredient in blood glucose meters used by diabetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hexacyanoferrate(III) is used in reading palimpsests and old manuscripts, in blueprint drawing and in photography (Cyanotype process). It finds application to harden iron and steel, in electroplating, dyeing wool and as a laboratory reagent. It is a mild oxidizing agent used in organic synthesis. It is associated with potassium hydroxide solution used to formulate Murakami’s etchant, which is useful for metallographers to provide contrast between binder and carbide phases in cemented carbides. Further, it is used in many amperometric biosensors as an electron transfer agent replacing an enzyme′s natural electron transfer agent such as oxygen with the enzyme glucose oxidase. It is an active ingredient in blood glucose meters used by diabetics.
Solubility
Soluble in water and acid. Slightly soluble in alcohol.
Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ammonia, hydrochloric acid and cyanides.
Specifications
Specifications
| Melting Point | (decomposition) |
| Quantity | 5000 g |
| Linear Formula | K3Fe(CN)6 |
| Merck Index | 14,7630 |
| Solubility Information | Soluble in water and acid. Slightly soluble in alcohol. |
| IUPAC Name | tripotassium hexacyanoirontris(ylium) |
| Formula Weight | 329.26 |
| Percent Purity | ≥98% |
| Sensitivity | Light sensitive |
| Density | 1.89 |
| Show More |
Safety and Handling
| EINECSNumber | 237-323-3 |
| RTECSNumber | LJ8225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.