Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hexafluorophosphate 95.0+%, TCI America™
SDP

Catalog No. P1023100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1023100G 100 g
P102325G 25 g
P1023500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1023100G Supplier TCI America Supplier No. P1023100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
Color White
pH 3.0 to 7.0 (50g/L H2O soln.)
Physical Form Crystalline Powder
Quantity 100 g
UN Number 3260
Formula Weight 184.06
Percent Purity ≥95.0% (T)
Chemical Name or Material Potassium Hexafluorophosphate
EINECSNumber (1)-0323
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.