Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Diiodate, ≥99.8% (RT), Solstice

Catalog No. 60047176 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-176 500 g Glass bottle
60-047-175 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-176 Supplier Solstice Supplier No. 60350500G

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.8% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-24-8
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
MDL Number MFCD00011400
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Linear Formula KH(IO3)2
UN Number UN3085
Formula Weight 389.91g/mol
Percent Purity ≥99.8% (RT)
Packaging Glass bottle
Chemical Name or Material Potassium hydrogen diiodate
Hazard Statement P220-P280-P305 + P351 + P338-P310
DOTInformation 8(5.1) / PGII
RTECSNumber 236-650-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.