CAS RN 13455-24-8
Potassium hydrogen iodate, 0.025N Standardized Solution
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 MDL Number: MFCD00011400 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 MDL Number: MFCD00011400 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, 0.0282 N (0.00235 M), 1000 ppm as Cl2, Ricca Chemical
1 mL = 1 mg Cl2, ASTM for Chlorine Testing
Potassium Biiodate, 0.0250 N, 0.002083 M, Ricca Chemical
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, 0.0375 N, Ricca Chemical
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, 0.00564 N, 200 ppm as Cl2, Ricca Chemical
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, 0.100 N (N/10), Ricca Chemical
CAS: 13455-24-8 Molecular Formula: HI2KO6 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M
Potassium Dihydrogen Diiodate, ACS Reagent Grade, Ricca Chemical
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, Certified, 0.0250N ±0.0005N (0.0021M), LabChem™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, ACS Grade, 99.95 to 100.05%, LabChem™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, For Residual Chlorine, Certified, 0.0282N ±0.0005N (0.00235M), LabChem™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, Certified, 0.0375N ±0.0005N (0.00313M), LabChem™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Biiodate, Certified, 0.1000N ±0.0005N (0.0085M), LabChem™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]
Potassium Bi-iodate,0.100 N, AquaPhoenix Scientific™
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.91 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M IUPAC Name: potassium iodic acid iodate SMILES: [K+].O[I](=O)=O.[O-][I](=O)=O
Potassium Hydrogen Diiodate, ≥99.8% (RT), Solstice
CAS: 13455-24-8 Molecular Formula: HI2KO6 Molecular Weight (g/mol): 389.909 MDL Number: MFCD00011400 InChI Key: ACAYDTMSDROWHW-UHFFFAOYSA-M Synonym: potassium hydrogen diiodate,potassium biiodate,kaliumhydrogendijodat,iodic acid hio3 , potassium salt 2:1,potassium biiodate solution,potassium iodic acid iodate,io3.k.hio3,potassium ion iodic acid iodate,potassium hydrogen iodate powder,potassium hydrogen diiodate, p.a PubChem CID: 23700942 IUPAC Name: potassium;iodic acid;iodate SMILES: OI(=O)=O.[O-]I(=O)=O.[K+]