Learn More
Potassium Hydrogen Phthalate, 8000 mg/L, Reagents
Specifications
Specifications
| pH | 4 |
| Boiling Point | 100°C |
| Color | Colorless |
| Quantity | 100 mL |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.004 |
| Grade | Reagent |
| Chemical Name or Material | Potassium Hydrogen Phthalate, 8000 mg/L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.