Learn More
Potassium Hydrogen Phthalate, 99.95 to 100.05%, ACS, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Color | White |
| Physical Form | Crystals |
| Quantity | 125 g |
| Assay Percent Range | 99.95 to 100.05% |
| Density | 1.636g/cm3; Bulk Density: 900 kg/m3 |
| Formula Weight | 204.22 |
| Chemical Name or Material | Potassium Hydrogen Phthalate, Secondary Reference Material For Alkalimetry, Traceable To NIST Standard Reference Material (SRM) Certipure™ |
Safety and Handling
| Recommended Storage | Store at +15°C to +25°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.