Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen phthalate, 99%

Catalog No. AAA128950I
Change view
Click to view available options
Quantity:
500 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA128950I 5000 g
AAA1289536 500 g
AAA128950B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA128950I Supplier Thermo Scientific Chemicals Supplier No. A128950I
Only null left
Add to Cart

CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol

Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 297°C (decomposition)
Density 1.636
Odor Odorless
Quantity 5000 g
Beilstein 3637128
Merck Index 14,7612
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 204.23
Percent Purity 99%
Chemical Name or Material Potassium hydrogen phthalate
EINECSNumber 212-889-4
RTECSNumber CZ4326000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.