Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen phthalate, for HPLC

Catalog No. AC463852500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463852500 250 g
AC463850500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463852500 Supplier Thermo Scientific Chemicals Supplier No. 463852500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
IUPAC Name potassium 2-carboxybenzoate
SMILES [K+].OC(=O)C1=CC=CC=C1C([O-])=O

Specifications

Melting Point 295.0°C to 300.0°C
Quantity 250 g
Linear Formula 2-(HO2C)C6H4CO2K
Solubility Information Solubility aq. soln.: 80 g/L (20°C). Other solubilities: 330 g/L water (100°C), slightly soluble in alcohol
Formula Weight 204.22
Grade HPLC
Chemical Name or Material Potassium hydrogen phthalate
EINECSNumber 212-889-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.