Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Phthalate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCP0480500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP0480500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCP0480500 Supplier Ricca Chemical Company Supplier No. RDCP0480500B1
Add to Cart
Add to Cart

Potassium Acid Phthalate

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 500 g
Percent Purity 99.95 to 100.05%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Potassium Hydrogen Phthalate
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.