Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Phthalate 99.8+%, TCI America™
SDP

Catalog No. P030925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030925G 25 g
P0309500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P030925G Supplier TCI America Supplier No. P030925G
Add to Cart
Add to Cart

Specifications

CAS 877-24-7
Color White
pH 4.0 (0.05M H2O soln. 25°C)
Physical Form Crystalline
Quantity 25 g
Synonym Monopotassium Phthalate, Phthalic Acid Monopotassium Salt
Molecular Weight (g/mol) 204.222
Percent Purity ≥99.8% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.