Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.0500 N (N/20), Ricca Chemical
SDP

Catalog No. 619832 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6198-32 1 L Natural Poly Bottle
6198-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6198-32 Supplier Ricca Chemical Company Supplier No. 619832
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-58-3 | HKO | 56.11 g/mol

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11 g/mol
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 98%, 0.32
CAS Max % 100%, 0.33
Concentration or Composition Notes 0.0500Normal
Quantity 1 L
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.