Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.100 N In Isopropyl Alcohol, Reagents

Catalog No. 01334326
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-326 1 L
01-334-325 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-326 Supplier Reagents Holdings Llc Supplier No. CP1177001A
Add to Cart
Add to Cart

Potassium Hydroxide, 0.100 N In Isopropyl Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 1310-58-3 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 13
Color Colorless
Boiling Point 82.5°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.7861
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.100 N In Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.