Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.5 N Standardized, Reagents

Catalog No. 01334319
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-334-319 4 L Poly Bottle
01-334-322 10 L Cubitainer
01-334-321 1 L Poly Bottle
01-334-320 20 L Cubitainer
01-334-318 4 L Cubitainer
01-334-317 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-334-319 Supplier Reagents Holdings Llc Supplier No. CP1193004A
Add to Cart
Add to Cart

Potassium Hydroxide, 0.5 N Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Hydroxide, 0.5 N Standardized
pH 13.7
Color Colorless
CAS 7732-18-5, 1310-58-3
Boiling Point 100°C
Molecular Formula KOH/H2O
Synonym Caustic Potash, Potash Lye
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0236
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.