Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 1.0 N, Reagents

Catalog No. 01334332
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-332 4 L
01-334-334 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-332 Supplier Reagents Holdings Llc Supplier No. CP1164004A
Only null left
Add to Cart
Add to Cart

Potassium Hydroxide, 1.0 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name water
SMILES O

Specifications

pH 14
Color Colorless
Boiling Point 101°C
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0324
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 1.0 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.