Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 10.0 N, Ricca Chemical
SDP

Catalog No. 62529032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
30 gal.
Packaging:
Natural Poly Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
62529032 1 L Natural Poly Bottle
6252-901 4 L Natural Poly Bottle
6252-903 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 62529032 Supplier Ricca Chemical Company Supplier No. 6252.9032
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 60.0, 38.44
CAS Max % 62.0, 40.01
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10.0N
Chemical Name or Material Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.