Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution (10%), ENG Scientific

Catalog No. ES9779
Change view
Click to view available options
Quantity:
1 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9779 1 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES9779 Supplier Eng Scientific Inc Supplier No. 9779
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Hydroxide Solution (10%)

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Weight (g/mol) 56.11

Specifications

Quantity 1 x 100 mL
Percent Purity 10%
Packaging Bottle
Chemical Name or Material Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.