Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 10% (w/v), Solution, Spectrum™ Chemical
SDP

Catalog No. 18612736 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-736 4 L Poly Bottle
18-612-702 500 mL Poly Bottle
18-612-735 1 L Poly Bottle
18-612-703 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-736 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P3774LTPL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-58-3

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Concentration 10% ± 0.3%
Quantity 4 L
Percent Purity 10% ±0.3%
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.