Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 4.00 N, Ricca Chemical
SDP

Catalog No. 62525 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
62525 20 L Cubitainer
R6252000500 500 mL Natural Poly Bottle
R62520001A 1 L Natural Poly Bottle
R62520004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 62525 Supplier Ricca Chemical Company Supplier No. 62525
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH >13
Color Colorless
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
CAS 7732-18-5
Concentration or Composition Notes 4.00Normal
Quantity 20 L
Grade Laboratory
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.