Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Potassium hydroxide, 45% w/w aq. soln

Catalog No. AA46072AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46072AP 500 mL
AA46072K2 1 L
AA46072K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46072AP Supplier Thermo Scientific™ Supplier No. 46072AP
Add to Cart

CAS: 1310-58-3 | HKO | 56.11 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number UN1814
Density 1.456
Quantity 500 mL
Formula Weight 56.11
Physical Form Liquid
ShelfLife 3 Years
EINECSNumber 215-181-3
RTECSNumber TT2100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.