Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 45% (w/w) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18612715 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-715 200 L Poly Drum
18-612-712 500 mL Poly Bottle
18-612-713 4 L Poly Bottle
18-612-714 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-715 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1320200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-58-3 | 56.11 g/mol

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Quantity 200 L
Percent Purity 45%
Grade Reagent
Packaging Poly Drum
Chemical Name or Material Potassium Hydroxide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.