Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, 50% w/v aq. soln.

Catalog No. AA35621K2
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35621K2 1 L
AA35621AP 500 mL
AA35621K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35621K2 Supplier Thermo Scientific Chemicals Supplier No. 035621K2
  • $116.80
    $96.65 / Each of 1

    Save $20.15

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 1310-58-3 | HKO | 56.11 g/mol

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Density 1.5 g/cm3 at 20°C
Solubility Information Miscible with water and ethanol.
Quantity 1 L
Formula Weight 56.11
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Potassium hydroxide
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 215-181-3
RTECSNumber TT2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.