Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 50% (w/v), Suitable for Orsat Gas Analysis, Ricca Chemical
SDP

Catalog No. 616016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6160-16 500 mL Natural Poly Bottle
6160-32 1 L Natural Poly Bottle
6160-1 4 L Natural Poly Bottle
616055 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 6160-16 Supplier Ricca Chemical Company Supplier No. 616016
Add to Cart
Add to Cart
This item is not returnable. View return policy
Suitable for Orsat Gas Analysis

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Quantity 500 mL
Formula Weight 56.11
Percent Purity 49.94 to 50.06%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 50% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution, 50% (w/v)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.