Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 50% (w/v), Suitable for Orsat Gas Analysis, Ricca Chemical
SDP

Catalog No. 616055 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
616055 55 gal. Poly Drum
6160-16 500 mL Natural Poly Bottle
6160-32 1 L Natural Poly Bottle
6160-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 616055 Supplier Ricca Chemical Company Supplier No. 616055
Add to Cart
Add to Cart
Request a Quote
Suitable for Orsat Gas Analysis

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 50.0, 48.24
CAS Max % 52.0, 50.21
Concentration or Composition Notes 50% (w/v)
Quantity 55 gal.
Percent Purity 49.94 to 50.06%
Grade Laboratory
Packaging Poly Drum
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.