Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide ACS AR Pellets, Macron Fine Chemicals™

Catalog No. MK69841 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
POLYSTORMOR
Poly
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6984-1 1 kg POLYSTORMOR
MK-6984-212 2.5 kg Poly
MK-6984-500 500 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-6984-1 Supplier Avantor Supplier No. 698412
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Quantity 1 kg
Formula Weight 56.11
Grade ACS
Packaging POLYSTORMOR
Physical Form Pellet
Chemical Name or Material Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.