Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, ACS, 85%, Spectrum™ Chemical
SDP

Catalog No. 18612730 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
12 kg
50 kg
100 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-730 100 kg Fiber Drum
18-612-724 125 g Poly Bottle
18-612-725 500 g Poly Bottle
18-612-726 1 kg Poly Bottle
18-612-727 2.5 kg Poly Bottle
18-612-728 12 kg Poly Pail
18-612-729 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-612-730 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1315100KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

CAS Max % 1
Quantity 100 kg
Percent Purity 85%
Grade ACS
Identification To Pass Test
Packaging Fiber Drum

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.