Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, FCC, 85-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605600 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-600 50 kg Poly Bottle
18-605-595 500 g Poly Bottle
18-605-597 2.5 kg Poly Bottle
18-605-596 6 x 500 g Poly Bottle
18-605-598 4 x 2.5 kg Poly Bottle
18-605-599 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-600 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P131650KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1316, 1310-58-3, KOH

Spectrum™ Chemical Potassium Hydroxide, Pellets, FCC is used as a rinse or chemical peel for fruits and vegetables, a stabilizer to prolong shelf-life, and as a thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

CAS Max % 1
Physical Form Pellets
Quantity 50 kg
Percent Purity 85 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide, Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.