Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Solstice

Catalog No. 60045833 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-833 6 x 1 kg
60-012-72 500 g
60-045-834 6 x 500 g
60-012-70 1 kg
60-045-832 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-833 Supplier Solstice Supplier No. 306036X1KG
Add to Cart
Add to Cart

Puriss. p.a., Reag. Ph. Eur.,≥85%, pellets

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Boiling Point 1,327°C
Melting Point 361°C (literature)
CAS Min % ≥85.0000%
CAS Max % ≤100.0000%
Linear Formula KOH
UN Number UN1813
Density 2.04 g/cm3
Quantity 6 x 1 kg
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Percent Purity ≥85%
Grade puriss. p.a.
Packaging Poly Bottle
Physical Form Pellet
Chemical Name or Material Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Potassium hydroxide, solid
EINECSNumber 215-181-3
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.