Learn More
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Linear Formula | KOH |
| Concentration | 45% |
| UN Number | 1814 |
| Density | 1.450g/mL at 20°C |
| Quantity | 100 mL |
| IUPAC Name | potassium hydroxide |
| Formula Weight | 56.11g/mol |
| Grade | HPLC |
| Packaging | HDPE Bottle |
| Chemical Name or Material | Potassium Hydroxide Solution |
Safety and Handling
| Hazard Statement | P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310 |
| Precautionary Statement | H290-H302-H314 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.