Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, ACS Reagent, 99.5%, Solstice

Catalog No. 60045252 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-252 5 g Poly Bottle
60-045-247 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-252 Supplier Solstice Supplier No. 2159295G

ACS Reagent, 99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C
Color Colorless
pH 5 to 8 (at 20°C)
Physical Form Solid
Quantity 5 g
Linear Formula KIO3
UN Number UN1479
Formula Weight 214g/mol
Percent Purity 99.50%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Density 3.93g/mL at 25°C; Bulk Density: ca. 2400kg/cu.m
Chemical Name or Material Potassium Iodate
Show More Show Less
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H315-H319-H335
DOTInformation 5.1/PGII
EINECSNumber 231-831-9
RTECSNumber NN1350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.