Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical
SDP

Catalog No. 628025 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
62802-5 10 L Cubitainer
6280-16 500 mL Amber Glass Bottle
6280-32 1 L Amber Glass Bottle
6280-1 4 L Amber Glass Bottle
R62800004F 4 L Cubitainer
6280-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 62802-5 Supplier Ricca Chemical Company Supplier No. 62802.5
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS 7732-18-5, 7758-05-6, 7758-05-6, 1310-73-2
CAS Min % 98.0, 0.42, 0.05, 0.0016
CAS Max % 100.0, 0.44, 0.05, 0.0016
Synonym dihydrogen oxide, dihydrogen monoxide
Molecular Weight (g/mol) 214 g/mol
Equivalence 1 mL = 0.5 mg SO
Grade Laboratory
Packaging Cubitainer
pH Slightly Alkaline
Quantity 10 L
Physical Form Liquid
Concentration or Composition Notes 0.0125Normal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.