Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical
SDP

Catalog No. 62805 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6280-5 20 L Cubitainer
6280-16 500 mL Amber Glass Bottle
6280-32 1 L Amber Glass Bottle
6280-1 4 L Amber Glass Bottle
R62800004F 4 L Cubitainer
62802-5 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 6280-5 Supplier Ricca Chemical Company Supplier No. 62805
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Iodate-Iodide, 0.0125N, Ricca Chemical
CAS 7758-05-6, 7681-11-0, 1310-73-2, 7732-18-5
Molecular Formula IKO3
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
Equivalence 1 mL=0.5 mg SO32-
PubChem CID 23665710
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0125N
Packaging Cubitainer
pH Slightly Alkaline
Quantity 20 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.