Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 10% w/v With 0.3% Sodium Bicarbonate, Reagents

Catalog No. 01336722
Change view
Click to view available options
Quantity:
10 L
20 L
Packaging:
Cubitainer
Winpak
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-722 20 L Winpak
01-336-724 10 L Cubitainer
01-336-721 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-722 Supplier Reagents Holdings Llc Supplier No. C6317P0020E7
Add to Cart
Add to Cart

Potassium Iodide, 10% w/v With 0.3% Sodium Bicarbonate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless to Light Yellow
pH 7 to 8
Physical Form Liquid
Quantity 20 L
Percent Purity 10% w/v
Grade Reagent
Packaging Winpak
Specific Gravity 1.075
Chemical Name or Material Potassium Iodide, 10% w/v With 0.3% Sodium Bicarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.