Learn More
Potassium Iodide, 15% w/v, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless to Light Yellow |
| Physical Form | Liquid |
| Quantity | 4 L |
| Percent Purity | 15% |
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 1.1086 |
| Chemical Name or Material | Potassium Iodide, 15% w/v |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.