Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 5% (w/v), Stabilized, Ricca Chemical
SDP

Catalog No. 629016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6290-16 500 mL Amber Poly Bottle
6290-32 1 L Amber Poly Bottle
6290-1 4 L Black Poly Bottle
62902-5 10 L Cubitainer
62905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6290-16 Supplier Ricca Chemical Company Supplier No. 629016
Add to Cart
Add to Cart

APHA/ASTM/EPA for Chlorine Testing

Specifications

CAS 7732-18-5
Molecular Weight (g/mol) 166 g/mol
Percent Purity 5%
Grade Laboratory
Packaging Amber Poly Bottle
Methods APHA/ASTM/EPA for Chlorine
Quantity 500 mL
Chemical Name or Material Potassium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.