Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 50% w/v, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88690 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle,Orange Cap
Poly Bottle,Purple Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S88690 500 mL Poly Bottle,Orange Cap
S88691 500 mL Poly Bottle,Purple Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S88690 Supplier Aqua Phoenix Scientific Supplier No. PI1450P
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5 , 497-19-8
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 50% w/v
Packaging Poly Bottle,Orange Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.