Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Potassium iodide, 99%, for biochemistry

Catalog No. AC447111000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447111000 100 g
AC447115000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447111000 Supplier Thermo Scientific Chemicals Supplier No. 447111000
Add to Cart

CAS: 7681-11-0 | IK | 166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Quantity 100 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula KI
Merck Index 15,7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Percent Purity 99%
Grade Biochemical
Packaging Glass bottle
Chemical Name or Material Potassium iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-659-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.